Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pkma
Entrez Gene ID
335817
Gene Name
pyruvate kinase M1/2a
Gene Aliases
cb1029, fj33f03, pkm2, pkm2a, wu:fj33f03
Chromosome Location
Chr.18: 910938 - 951570 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199333.1 NP_955365.1 8 - 9 1200 93
GenBank mRNA BC045421.1 - 8 - 9 1200 93
BC164692.1 - 7 - 8 1179 93

View Full Product Details