Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
usp44
Entrez Gene ID
393227
Gene Name
ubiquitin specific peptidase 44
Gene Aliases
zgc:55661
Chromosome Location
Chr.4: 26021746 - 26035770 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200257.1 NP_956551.1 3 - 4 1563 100
GenBank mRNA BC048060.1 - 3 - 4 1563 100
BC164039.1 - 2 - 3 1479 100

View Full Product Details