Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pcmtl
Entrez Gene ID
393741
Gene Name
l-isoaspartyl protein carboxyl methyltransferase, like
Gene Aliases
zgc:73268
Chromosome Location
Chr.17: 981 - 8441 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200768.1 NP_957062.1 5 - 6 409 145
XM_005158766.4 XP_005158823.1 6 - 7 516 145
XM_005158767.4 XP_005158824.1 5 - 6 425 145
XM_009293125.3 XP_009291400.1 6 - 7 516 145
GenBank mRNA BC059599.1 - 5 - 6 409 145

View Full Product Details