Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
adrm1
Entrez Gene ID
796450
Gene Name
adhesion regulating molecule 1
Gene Aliases
adrm1a, adrm1b, fa12d09, wu:fa10g10, wu:fa12d09, zgc:56107, zgc:77256
Chromosome Location
Chr.23: 9560965 - 9567306 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201013.3 NP_957307.3 5 - 6 687 71
XM_005161962.4 XP_005162019.1 5 - 6 703 71
XM_021469854.1 XP_021325529.1 5 - 6 700 71
GenBank mRNA BC066391.1 - 5 - 6 666 71
BC154825.1 - 5 - 6 629 71
EH561147.1 - 5 - 6 827 71
EH594482.1 - 5 - 6 714 71

View Full Product Details