Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
adgrl4
Entrez Gene ID
406676
Gene Name
adhesion G protein-coupled receptor L4
Gene Aliases
eltd1, fd10a02, wu:fd10a02, zgc:63629
Chromosome Location
Chr.11: 7383213 - 7410314 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213367.2 NP_998532.2 6 - 7 740 126
GenBank mRNA BC055171.1 - 6 - 7 737 126

View Full Product Details