The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213367.2 | NP_998532.2 | 8 - 9 | 1036 | 78 |
GenBank mRNA | BC055171.1 | - | 8 - 9 | 1033 | 78 |