Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gtpbp10
Entrez Gene ID
436727
Gene Name
GTP-binding protein 10 (putative)
Gene Aliases
zgc:92334
Chromosome Location
Chr.16: 23891423 - 23902439 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002454.1 NP_001002454.1 5 - 6 616 84
GenBank mRNA BC076017.1 - 5 - 6 616 84

View Full Product Details