Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gtpbp10
Entrez Gene ID
436727
Gene Name
GTP-binding protein 10 (putative)
Gene Aliases
zgc:92334
Chromosome Location
Chr.16: 23891423 - 23902439 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002454.1 NP_001002454.1 7 - 8 771 90
GenBank mRNA BC076017.1 - 7 - 8 771 90

View Full Product Details