Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bmp2k
Entrez Gene ID
494101
Gene Name
BMP2 inducible kinase
Gene Aliases
zgc:101641
Chromosome Location
Chr.5: 436789 - 501728 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008644.2 NP_001008644.2 1 - 2 530 58
NM_001193540.1 NP_001180469.1 1 - 2 530 58
XM_005165303.4 XP_005165360.1 1 - 2 1175 58
GenBank mRNA BC086712.1 - 1 - 2 530 58

View Full Product Details