Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pfkmb
Entrez Gene ID
568001
Gene Name
phosphofructokinase, muscle b
Gene Aliases
im:7137110, wu:fc58g11
Chromosome Location
Chr.6: 306604 - 331891 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089417.2 NP_001082886.2 12 - 13 1264 142
NM_001365336.1 NP_001352265.1 15 - 16 1778 142
GenBank mRNA BC139532.1 - 12 - 13 1251 142
EB884905.1 - 5 - 6 380 142

View Full Product Details