Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pfkmb
Entrez Gene ID
568001
Gene Name
phosphofructokinase, muscle b
Gene Aliases
im:7137110, wu:fc58g11
Chromosome Location
Chr.6: 306604 - 331891 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089417.2 NP_001082886.2 1 - 2 159 72
NM_001365336.1 NP_001352265.1 4 - 5 673 72
GenBank mRNA BC139532.1 - 1 - 2 146 72
EB897876.1 - 1 - 2 171 72
EB899920.1 - 1 - 2 115 72

View Full Product Details