Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pfkmb
Entrez Gene ID
568001
Gene Name
phosphofructokinase, muscle b
Gene Aliases
im:7137110, wu:fc58g11
Chromosome Location
Chr.6: 306604 - 331891 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089417.2 NP_001082886.2 2 - 3 235 75
NM_001365336.1 NP_001352265.1 5 - 6 749 75
GenBank mRNA BC139532.1 - 2 - 3 222 75
EB897876.1 - 2 - 3 247 75
EB899920.1 - 2 - 3 191 75

View Full Product Details