Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myl7
Entrez Gene ID
30592
Gene Name
myosin, light chain 7, regulatory
Gene Aliases
cmlc2, mlc7, mylc2a, zgc:92755
Chromosome Location
Chr.8: 40457107 - 40464935 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131329.3 NP_571404.1 3 - 4 358 85
GenBank mRNA AF114428.1 - 1 - 2 72 85
AF425743.1 - 2 - 3 185 85
AY057074.1 - 3 - 4 227 85
BC076232.1 - 3 - 4 359 85

View Full Product Details