Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nitr4a
Entrez Gene ID
60652
Gene Name
novel immune-type receptor 4a
Gene Aliases
dZ105L16.2, dZ105L16.3, dZ105L16.8, nitr, nitr4a_9
Chromosome Location
Chr.7: 17578061 - 17582292 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131654.1 NP_571729.1 5 - 6 129
NM_131655.1 NP_571730.1 5 - 6 129
NM_131656.1 NP_571731.1 5 - 6 129
NM_131657.1 NP_571732.1 5 - 6 840 129
NM_131658.1 NP_571733.1 5 - 6 840 129
NM_198344.3 NP_938158.3 5 - 6 129
GenBank mRNA AF318402.1 - 5 - 6 129
AF318403.1 - 5 - 6 129
AF318404.1 - 5 - 6 129
AF318405.1 - 5 - 6 840 129
AF318406.1 - 5 - 6 840 129
CU638757.1 - 5 - 6 129

View Full Product Details