Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cryaa
Entrez Gene ID
100000769
Gene Name
crystallin, alpha A
Gene Aliases
wu:fc06h04, zgc:92036
Chromosome Location
Chr.1: 29096907 - 29100918 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152950.2 NP_694482.1 2 - 3 415 81
GenBank mRNA AY035778.1 - 2 - 3 404 81
BC083177.1 - 2 - 3 415 81
BC164886.1 - 2 - 3 344 81

View Full Product Details