Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zdhhc8b
Entrez Gene ID
352941
Gene Name
zinc finger DHHC-type palmitoyltransferase 8b
Gene Aliases
zdhhc5, zisp
Chromosome Location
Chr.5: 17881080 - 18007077 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178304.3 NP_840089.2 2 - 3 534 69
GenBank mRNA AY871211.1 - 2 - 3 534 69

View Full Product Details