Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gamt
Entrez Gene ID
796865
Gene Name
guanidinoacetate N-methyltransferase
Gene Aliases
zgc:123136
Chromosome Location
Chr.11: 62066 - 70533 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105595.1 NP_001099065.1 3 - 4 421 65
GenBank mRNA AF425745.1 - 3 - 4 444 65
BC107607.1 - 3 - 4 468 65
BC153601.1 - 3 - 4 421 65

View Full Product Details