Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crls1
Entrez Gene ID
405867
Gene Name
cardiolipin synthase 1
Gene Aliases
zgc:85909
Chromosome Location
Chr.17: 4368885 - 4386084 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212931.1 NP_998096.1 5 - 6 972 67
GenBank mRNA BC067687.1 - 5 - 6 972 67

View Full Product Details