Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
psmg2
Entrez Gene ID
573107
Gene Name
proteasome (prosome, macropain) assembly chaperone 2
Gene Aliases
tnfsf5ip1, wu:fb51b11, zgc:73278
Chromosome Location
Chr.19: 12350110 - 12360121 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213032.1 NP_998197.1 5 - 6 663 102
GenBank mRNA BC059607.1 - 5 - 6 663 102

View Full Product Details