Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
scinlb
Entrez Gene ID
406363
Gene Name
scinderin like b
Gene Aliases
wu:fe16f08, zgc:77481
Chromosome Location
Chr.2: 10396379 - 10450792 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213090.2 NP_998255.2 9 - 10 1251 61
GenBank mRNA BC066531.1 - 9 - 10 1216 61

View Full Product Details