Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pkmb
Entrez Gene ID
445094
Gene Name
pyruvate kinase M1/2b
Gene Aliases
pkm2b, zgc:92037
Chromosome Location
Chr.25: 29159817 - 29196072 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003488.1 NP_001003488.1 8 - 9 1323 93
XM_005163113.4 XP_005163170.3 9 - 10 1831 93
GenBank mRNA BC076497.1 - 8 - 9 1323 93
BC165710.1 - 8 - 9 1333 93

View Full Product Details