Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lmod2a
Entrez Gene ID
447877
Gene Name
leiomodin 2 (cardiac) a
Gene Aliases
zgc:103424
Chromosome Location
Chr.25: 27814309 - 27819944 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004616.2 NP_001004616.1 1 - 2 332 63
XM_005163115.3 XP_005163172.1 1 - 2 438 63
GenBank mRNA BC081475.1 - 1 - 2 334 63

View Full Product Details