Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rrh
Entrez Gene ID
447916
Gene Name
retinal pigment epithelium-derived rhodopsin homolog
Gene Aliases
zgc:103757
Chromosome Location
Chr.13: 12528118 - 12540539 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004654.1 NP_001004654.1 4 - 5 607 88
GenBank mRNA BC081524.1 - 4 - 5 607 88
BC164135.1 - 4 - 5 582 88
KT008425.1 - 4 - 5 548 88

View Full Product Details