Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp2ad3
Entrez Gene ID
569245
Gene Name
cytochrome P450, family 2, subfamily AD, polypeptide 3
Gene Aliases
cyp2j27, si:dkey-183n20.10
Chromosome Location
Chr.20: 25548425 - 25551676 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025554.2 NP_001020725.1 6 - 7 1057 104
GenBank mRNA AY500819.1 - 6 - 7 977 104
BC134877.1 - 6 - 7 1057 104

View Full Product Details