Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zte38
Entrez Gene ID
556133
Gene Name
zebrafish testis-expressed 38
Gene Aliases
zgc:136823
Chromosome Location
Chr.2: 6092065 - 6104015 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039809.1 NP_001034898.1 4 - 5 458 78
GenBank mRNA BC114280.1 - 4 - 5 458 78

View Full Product Details