Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pofut2
Entrez Gene ID
777668
Gene Name
protein O-fucosyltransferase 2
Gene Aliases
fc46a11, wu:fc46a11, zgc:194822
Chromosome Location
Chr.9: 40850988 - 40873967 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077630.2 NP_001071098.2 8 - 9 1232 83
GenBank mRNA BC163087.1 - 8 - 9 1232 83
BC163097.1 - 8 - 9 1232 83
DQ139956.1 - 8 - 9 1180 83

View Full Product Details