Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
scn3b
Entrez Gene ID
563935
Gene Name
sodium channel, voltage-gated, type III, beta
Gene Aliases
-
Chromosome Location
Chr.15: 22133443 - 22147873 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080802.1 NP_001074271.1 5 - 6 947 79
XM_009291661.2 XP_009289936.1 5 - 6 959 79
GenBank mRNA BC163285.1 - 5 - 6 844 79
BC163295.1 - 5 - 6 844 79
DQ489730.1 - 5 - 6 947 79

View Full Product Details