Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tspan4b
Entrez Gene ID
556934
Gene Name
tetraspanin 4b
Gene Aliases
tspan4, zgc:163023
Chromosome Location
Chr.3: 73905 - 83364 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082807.1 NP_001076276.1 6 - 7 1042 85
GenBank mRNA BC134198.1 - 6 - 7 1042 85

View Full Product Details