Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aga
Entrez Gene ID
566517
Gene Name
aspartylglucosaminidase
Gene Aliases
zgc:77327
Chromosome Location
Chr.14: 204941 - 214062 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110281.1 NP_001103751.1 8 - 9 960 109
XM_021481042.1 XP_021336717.1 6 - 7 675 109
GenBank mRNA BC154306.1 - 8 - 9 960 109

View Full Product Details