Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nipsnap1
Entrez Gene ID
30231
Gene Name
nipsnap homolog 1 (C. elegans)
Gene Aliases
zgc:153339
Chromosome Location
Chr.5: 12032802 - 12063377 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131033.2 NP_571108.2 8 - 9 984 91
GenBank mRNA BC122249.1 - 8 - 9 984 91
EH486911.1 - 2 - 3 513 91
EH513303.1 - 8 - 9 743 91

View Full Product Details