Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atp6v1ba
Entrez Gene ID
359839
Gene Name
ATPase, H+ transporting, lysosomal, V1 subunit B, member a
Gene Aliases
cb927, fb52g11, vatB1, wu:fb52g11
Chromosome Location
Chr.17: 49201 - 95408 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182878.1 NP_878298.1 12 - 13 1332 77
GenBank mRNA AF472614.1 - 12 - 13 1332 77
BC052222.1 - 12 - 13 1290 77
BC059455.1 - 12 - 13 1282 77
BC071387.1 - 12 - 13 1330 77

View Full Product Details