Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tardbpl
Entrez Gene ID
386630
Gene Name
TAR DNA binding protein, like
Gene Aliases
tardbp, wu:fb52e05, wu:fe47c11, wu:fj94b04
Chromosome Location
Chr.23: 29357753 - 29362846 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001362353.1 NP_001349282.1 2 - 3 800 122
NM_198364.2 NP_938178.1 2 - 3 800 122
XM_021469761.1 XP_021325436.1 2 - 3 868 122
XM_021469762.1 XP_021325437.1 2 - 3 868 122
GenBank mRNA BC044405.1 - 2 - 3 669 122
BC164253.1 - 2 - 3 440 122
EH537260.1 - 2 - 3 288 122

View Full Product Details