Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tardbpl
Entrez Gene ID
386630
Gene Name
TAR DNA binding protein, like
Gene Aliases
tardbp, wu:fb52e05, wu:fe47c11, wu:fj94b04
Chromosome Location
Chr.23: 29357753 - 29362846 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001362353.1 NP_001349282.1 6 - 7 1341 142
NM_198364.2 NP_938178.1 6 - 7 1356 142
GenBank mRNA BC044405.1 - 6 - 7 1225 142
BC066430.1 - 6 - 7 1341 142
EH537260.1 - 6 - 7 829 143

View Full Product Details