Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pdcd4b
Entrez Gene ID
321061
Gene Name
programmed cell death 4b
Gene Aliases
cb45, pdcd4, sb:cb45, wu:fc84b08
Chromosome Location
Chr.22: 29658983 - 29689999 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198978.1 NP_945329.1 3 - 4 497 117
XM_005161791.4 XP_005161848.1 4 - 5 580 117
GenBank mRNA BC045513.1 - 3 - 4 497 117
BC065341.1 - 4 - 5 522 117

Gene Symbol
LOC103909982
Entrez Gene ID
103909982
Gene Name
programmed cell death protein 4-like
Gene Aliases
-
Chromosome Location
Chr.Unknown: 1709 - 6180 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_009298358.3 XP_009296633.1 1 - 2 132 117

View Full Product Details