Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdcd4b
Entrez Gene ID
321061
Gene Name
programmed cell death 4b
Gene Aliases
cb45, pdcd4, sb:cb45, wu:fc84b08
Chromosome Location
Chr.22: 29658983 - 29689999 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198978.1 NP_945329.1 7 - 8 1042 125
XM_005161791.4 XP_005161848.1 8 - 9 1125 125
GenBank mRNA BC045513.1 - 7 - 8 1042 125
BC065341.1 - 8 - 9 1067 125

View Full Product Details