Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eapp
Entrez Gene ID
327318
Gene Name
e2f-associated phosphoprotein
Gene Aliases
c14orf11, wu:fd21g02, zgc:65820
Chromosome Location
Chr.17: 9951618 - 9962527 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199784.1 NP_956078.1 4 - 5 553 63
GenBank mRNA BC056728.1 - 4 - 5 553 63

View Full Product Details