The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201489.1 | NP_958897.1 | 2 - 3 | 254 | 114 |
GenBank mRNA | BC049139.1 | - | 2 - 3 | 254 | 114 |
BC165255.1 | - | 2 - 3 | 168 | 114 |