Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cacna1da
Entrez Gene ID
403310
Gene Name
calcium channel, voltage-dependent, L type, alpha 1D subunit, a
Gene Aliases
cacna1d, cav1.3a, isl, si:dz136d19.3, si:dz223c6.1
Chromosome Location
Chr.11: 36792831 - 36964176 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203484.1 NP_982351.1 44 - 45 5598 74
XM_021479576.1 XP_021335251.1 45 - 46 5819 74
XM_021479577.1 XP_021335252.1 45 - 46 5819 74
XM_021479578.1 XP_021335253.1 45 - 46 5816 74
XM_021479579.1 XP_021335254.1 44 - 45 5792 74
XM_021479580.1 XP_021335255.1 44 - 45 5785 74
XM_021479581.1 XP_021335256.1 44 - 45 5743 74
XM_021479582.1 XP_021335257.1 43 - 44 5438 74
XM_021479583.1 XP_021335258.1 44 - 45 5702 74
XM_021479584.1 XP_021335259.1 43 - 44 5710 74
GenBank mRNA AY528224.1 - 44 - 45 5598 74
BC163679.1 - 43 - 44 5438 74

View Full Product Details