Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
midn
Entrez Gene ID
323713
Gene Name
midnolin
Gene Aliases
fc07f11, wu:fc07f11, zgc:77423
Chromosome Location
Chr.22: 18724992 - 18749524 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207052.1 NP_996935.1 5 - 6 1254 67
XM_005170436.4 XP_005170493.1 6 - 7 1242 67
XM_005170437.4 XP_005170494.1 5 - 6 1010 67
GenBank mRNA BC066456.1 - 5 - 6 1254 67

View Full Product Details