Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnajc4
Entrez Gene ID
324373
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 4
Gene Aliases
fa66f02, wu:fa66f02, wu:fc26h09, zgc:56236, zgc:77513
Chromosome Location
Chr.14: 103098 - 123349 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212677.1 NP_997842.1 6 - 7 678 81
GenBank mRNA BC047848.1 - 6 - 7 678 81
BC067159.1 - 6 - 7 670 81

View Full Product Details