Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnot3a
Entrez Gene ID
449540
Gene Name
CCR4-NOT transcription complex, subunit 3a
Gene Aliases
cnot3, fb73b06, fc17h07, wu:fb73b06, wu:fc17h07, zgc:92813
Chromosome Location
Chr.16: 32110195 - 32128370 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328118.1 NP_001315047.1 13 - 14 2224 61
XM_005158349.4 XP_005158406.1 13 - 14 2228 61
XM_021466678.1 XP_021322353.1 13 - 14 2243 61
GenBank mRNA BC081678.1 - 10 - 11 1274 61

View Full Product Details