Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnot3a
Entrez Gene ID
449540
Gene Name
CCR4-NOT transcription complex, subunit 3a
Gene Aliases
cnot3, fb73b06, fc17h07, wu:fb73b06, wu:fc17h07, zgc:92813
Chromosome Location
Chr.16: 32110195 - 32128370 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328118.1 NP_001315047.1 7 - 8 645 90
XM_005158349.4 XP_005158406.1 7 - 8 670 90
GenBank mRNA BC081678.1 - 6 - 7 523 90

View Full Product Details