Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fam50a
Entrez Gene ID
550329
Gene Name
family with sequence similarity 50 member A
Gene Aliases
db03h09, mg:db03h09, wu:ft29b08, zgc:110236
Chromosome Location
Chr.23: 33987382 - 34005296 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017636.1 NP_001017636.1 11 - 12 1073 76
GenBank mRNA BC092815.1 - 11 - 12 1073 76

View Full Product Details