Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mstna
Entrez Gene ID
445816
Gene Name
myostatin a
Gene Aliases
gdf8l, mstn-2, mstn2, si:dkeyp-84a8.5
Chromosome Location
Chr.22: 12790758 - 12795471 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY614000.2 - 3 - 3 1180 67

View Full Product Details