Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glt8d2
Entrez Gene ID
567942
Gene Name
glycosyltransferase 8 domain containing 2
Gene Aliases
si:dkey-22l11.1, zgc:136873
Chromosome Location
Chr.4: 268133 - 276433 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039931.1 NP_001035020.1 7 - 8 811 69
GenBank mRNA BC114297.1 - 7 - 8 811 69

View Full Product Details