Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
isy1
Entrez Gene ID
570205
Gene Name
ISY1 splicing factor homolog
Gene Aliases
im:7159238, wu:fc23h01, wu:fi21h07, zgc:136785
Chromosome Location
Chr.11: 474439 - 486306 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083563.2 NP_001077032.1 6 - 7 542 77
GenBank mRNA BC115265.1 - 6 - 7 316 77

View Full Product Details