Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hoxc3a
Entrez Gene ID
30421
Gene Name
homeobox C3a
Gene Aliases
HOX-114, zgc:110503
Chromosome Location
Chr.23: 36086681 - 36149044 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134685.2 NP_001128157.2 4 - 5 592 69
XM_005162365.4 XP_005162422.1 4 - 5 899 69
XM_005162366.4 XP_005162423.1 4 - 5 618 69
XM_005162367.4 XP_005162424.1 4 - 5 527 69
XM_021469727.1 XP_021325402.1 3 - 4 1156 69
GenBank mRNA BC095299.1 - 4 - 5 577 69
BC154496.1 - 4 - 5 592 69
BC165158.1 - 2 - 3 422 69
DQ060551.1 - 1 - 2 244 69
X60095.1 - - 1153 69

View Full Product Details