Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
foxd3
Entrez Gene ID
30548
Gene Name
forkhead box D3
Gene Aliases
fkd-6, fkd6, fkh6, forkhead-6, zgc:111934
Chromosome Location
Chr.6: 32092032 - 32093830 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131290.2 NP_571365.2 1 - 1 1187 141
GenBank mRNA AF052249.1 - 1 - 1 1152 141
BC095603.1 - 1 - 1 1187 141
EH608338.1 - 1 - 1 503 141

View Full Product Details