Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
psme1
Entrez Gene ID
30648
Gene Name
proteasome activator subunit 1
Gene Aliases
fk89d05, wu:fk89d05
Chromosome Location
Chr.24: 12945869 - 12954482 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131375.2 NP_571450.1 5 - 6 439 158
GenBank mRNA AF195052.1 - 5 - 6 440 158
BC075864.1 - 5 - 6 158

View Full Product Details