Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxw4
Entrez Gene ID
58035
Gene Name
F-box and WD repeat domain containing 4
Gene Aliases
dac, fk63g06, hag, hagoromo, wu:fk63g06
Chromosome Location
Chr.13: 28512877 - 28561512 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131521.2 NP_571596.1 8 - 9 1270 60
GenBank mRNA AB022162.1 - 8 - 9 1272 60
BC066481.1 - 8 - 9 1174 60

View Full Product Details